Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRINP2
Entrez Gene ID
709377
Gene Name
BMP/retinoic acid inducible neural specific 2
Gene Aliases
EGK_01993, FAM5B
Chromosome Location
Chr.1: 34408867 - 34527989 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265618.1 NP_001252547.1 - 586 130
XM_028843587.1 XP_028699420.1 - 588 130
XM_028843589.1 XP_028699422.1 - 744 130
XM_028843593.1 XP_028699426.1 - 422 130
GenBank mRNA JU321881.1 - - 586 130
JV667437.1 - - 586 130

View Full Product Details