Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HES4
Entrez Gene ID
698434
Gene Name
hes family bHLH transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.1: 223419527 - 223424359 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001090404.4 XP_001090404.2 - 1867 120
GenBank mRNA JV589257.1 - - 267 120

View Full Product Details