Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KMT2C
Entrez Gene ID
716753
Gene Name
lysine methyltransferase 2C
Gene Aliases
MLL3
Chromosome Location
Chr.3: 178118161 - 178416058 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015135341.2 XP_014990827.2 - 14397 162
XM_015135342.2 XP_014990828.2 - 13788 162
XM_028846497.1 XP_028702330.1 - 14652 162
XM_028846498.1 XP_028702331.1 - 14594 162
XM_028846499.1 XP_028702332.1 - 14591 162
XM_028846500.1 XP_028702333.1 - 14591 162
XM_028846501.1 XP_028702334.1 - 14582 162
XM_028846502.1 XP_028702335.1 - 14561 162
XM_028846503.1 XP_028702336.1 - 14429 162
XM_028846504.1 XP_028702337.1 - 14879 162
XM_028846505.1 XP_028702338.1 - 14350 162
XM_028846506.1 XP_028702339.1 - 14313 162
XM_028846507.1 XP_028702340.1 - 14726 162
GenBank mRNA JV589493.1 - - 2663 162
JV589494.1 - - 2651 162

View Full Product Details