Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF2L1
Entrez Gene ID
697928
Gene Name
stromal cell derived factor 2 like 1
Gene Aliases
-
Chromosome Location
Chr.10: 31302642 - 31304469 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145067.1 NP_001138539.1 - 384 100
GenBank mRNA FJ607063.1 - - 384 100
JU473505.1 - - 405 100

View Full Product Details