Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTARC1
Entrez Gene ID
705425
Gene Name
mitochondrial amidoxime reducing component 1
Gene Aliases
MARC1, MOSC1
Chromosome Location
Chr.1: 78397050 - 78440403 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102284.3 XP_001102284.1 - 2493 87
GenBank mRNA JU323318.1 - - 87

View Full Product Details