Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC86
Entrez Gene ID
695978
Gene Name
coiled-coil domain containing 86
Gene Aliases
EGK_06160
Chromosome Location
Chr.14: 13380764 - 13389172 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265928.1 NP_001252857.1 - 825 111
GenBank mRNA JU336346.1 - - 811 111
JV635881.1 - - 825 111
JV723193.1 - - 210 111

View Full Product Details