Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF23
Entrez Gene ID
714538
Gene Name
PHD finger protein 23
Gene Aliases
EGK_08108
Chromosome Location
Chr.16: 7002525 - 7007014 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106748.4 XP_001106748.1 - 281 80
GenBank mRNA JU336355.1 - - 166 80
JU477175.1 - - 174 80
JV048231.1 - - 197 80
JV661222.1 - - 197 80

View Full Product Details