Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LENG1
Entrez Gene ID
693417
Gene Name
leukocyte receptor cluster member 1
Gene Aliases
-
Chromosome Location
Chr.19: 53913500 - 53917681 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082125.4 XP_001082125.3 - 424 88
GenBank mRNA JU493317.1 - - 254 88

View Full Product Details