Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC31
Entrez Gene ID
697710
Gene Name
leucine rich repeat containing 31
Gene Aliases
-
Chromosome Location
Chr.2: 74768284 - 74799925 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015131644.2 XP_014987130.2 - 1281 118

View Full Product Details