Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RERGL
Entrez Gene ID
702819
Gene Name
RERG like
Gene Aliases
EGK_03437
Chromosome Location
Chr.11: 19478981 - 19489209 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095306.4 XP_001095306.3 - 479 152
XM_015151209.2 XP_015006695.2 - 322 152
GenBank mRNA JU326457.1 - - 55 152
JV071549.1 - - 97 152

View Full Product Details