Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT18
Entrez Gene ID
707061
Gene Name
nudix hydrolase 18
Gene Aliases
-
Chromosome Location
Chr.8: 22271754 - 22273746 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190895.1 NP_001177824.1 - 380 61

View Full Product Details