Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHS1
Entrez Gene ID
703396
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
-
Chromosome Location
Chr.9: 114262242 - 114310598 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015148216.2 XP_015003702.2 - 2482 100
XM_015148217.2 XP_015003703.2 - 2482 100
XM_028826883.1 XP_028682716.1 - 2478 100

View Full Product Details