Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAA50
Entrez Gene ID
709764
Gene Name
N-alpha-acetyltransferase 50, NatE catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.2: 162511203 - 162538312 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261371.2 NP_001248300.1 - 462 112
XM_015129813.2 XP_014985299.1 - 611 112
XM_028843327.1 XP_028699160.1 - 742 112
XM_028843328.1 XP_028699161.1 - 739 112
GenBank mRNA JU336491.1 - - 565 112
JU471806.1 - - 471 112
JV043838.1 - - 464 112
JV637198.1 - - 565 112
JV697188.1 - - 567 112

View Full Product Details