Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COQ10B
Entrez Gene ID
697432
Gene Name
coenzyme Q10B
Gene Aliases
EGK_04652
Chromosome Location
Chr.12: 84547096 - 84583717 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195786.1 NP_001182715.1 - 117 165
XM_028830416.1 XP_028686249.1 - 2090 165
GenBank mRNA EG703544.1 - - 97 165
JU336492.1 - - 172 165
JV048307.1 - - 151 165

View Full Product Details