Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRPEL1
Entrez Gene ID
709772
Gene Name
GrpE like 1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.5: 5900507 - 5920255 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261551.1 NP_001248480.1 - 91 68
XM_028848093.1 XP_028703926.1 - 250 68
XM_028848094.1 XP_028703927.1 - 250 68
GenBank mRNA JU320862.1 - - 91 68
JV602063.1 - - 76 68
JV662210.1 - - 88 68

View Full Product Details