Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT3
Entrez Gene ID
716909
Gene Name
Wnt family member 3
Gene Aliases
-
Chromosome Location
Chr.16: 57171387 - 57225633 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028836978.1 XP_028692811.1 - 744 114
GenBank mRNA JV070447.1 - - 181 114
JV660593.1 - - 159 114

View Full Product Details