Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATS1
Entrez Gene ID
703218
Gene Name
spermatogenesis associated serine rich 1
Gene Aliases
EGK_14962
Chromosome Location
Chr.4: 124953016 - 124986767 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028847503.1 XP_028703336.1 - 256 117
GenBank mRNA JV460293.1 - - 122 117

View Full Product Details