Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISL2
Entrez Gene ID
707916
Gene Name
ISL LIM homeobox 2
Gene Aliases
-
Chromosome Location
Chr.7: 53547254 - 53552996 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001105068.4 XP_001105068.1 - 1128 70

View Full Product Details