Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CERS5
Entrez Gene ID
713449
Gene Name
ceramide synthase 5
Gene Aliases
LASS5
Chromosome Location
Chr.11: 49738896 - 49775474 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102810.4 XP_001102810.3 - 728 146
XM_015151690.2 XP_015007176.2 - 668 146
XM_015151691.2 XP_015007177.2 - 668 146
XM_015151692.2 XP_015007178.1 - 668 146
XM_028829467.1 XP_028685300.1 - 668 146
XM_028829468.1 XP_028685301.1 - 668 146
GenBank mRNA JU337024.1 - - 455 146
JV075212.1 - - 388 146

View Full Product Details