Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMBX1
Entrez Gene ID
709514
Gene Name
diencephalon/mesencephalon homeobox 1
Gene Aliases
EGK_00718
Chromosome Location
Chr.1: 177889026 - 177894909 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015138807.2 XP_014994293.1 - 689 142

View Full Product Details