Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM219A
Entrez Gene ID
700651
Gene Name
family with sequence similarity 219 member A
Gene Aliases
C15H9orf25, C9orf25
Chromosome Location
Chr.15: 49741550 - 49801019 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193839.2 NP_001180768.1 - 562 105
XM_002800103.3 XP_002800149.1 - 546 105
XM_015117356.2 XP_014972842.1 - 597 105
XM_015117357.2 XP_014972843.1 - 546 105
GenBank mRNA JU322134.1 - - 515 105
JU322135.1 - - 515 105
JU322219.1 - - 566 105
JU472014.1 - - 498 105
JV601564.1 - - 566 105

View Full Product Details