Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX26
Entrez Gene ID
713363
Gene Name
testis expressed 26
Gene Aliases
C13orf26
Chromosome Location
Chr.17: 9980099 - 10023307 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194161.1 NP_001181090.1 - 824 157
XM_015120823.2 XP_014976309.1 - 821 157
XM_028837156.1 XP_028692989.1 - 765 157
XM_028837157.1 XP_028692990.1 - 971 157
XM_028837158.1 XP_028692991.1 - 599 157
XM_028837159.1 XP_028692992.1 - 681 157

View Full Product Details