Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCL2
Entrez Gene ID
696014
Gene Name
phosducin like 2
Gene Aliases
-
Chromosome Location
Chr.5: 78041582 - 78075992 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001087954.4 XP_001087954.1 - 497 127

View Full Product Details