Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRFN5
Entrez Gene ID
700512
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Gene Aliases
-
Chromosome Location
Chr.7: 103739129 - 104023244 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028851518.1 XP_028707351.1 - 1717 133

View Full Product Details