Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R21
Entrez Gene ID
715917
Gene Name
protein phosphatase 1 regulatory subunit 21
Gene Aliases
EGK_05315, KLRAQ1
Chromosome Location
Chr.13: 60230960 - 60323275 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112318.2 XP_014967804.1 - 1398 98
XM_015112321.2 XP_014967807.1 - 1434 98
XM_028831687.1 XP_028687520.1 - 1247 98
XM_028831688.1 XP_028687521.1 - 1381 98
XM_028831689.1 XP_028687522.1 - 1243 98
XM_028831690.1 XP_028687523.1 - 1420 98
XM_028831691.1 XP_028687524.1 - 1187 98
GenBank mRNA JU332921.1 - - 1330 98
JU474323.1 - - 1288 98
JV706950.1 - - 1352 98

View Full Product Details