Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R21
Entrez Gene ID
715917
Gene Name
protein phosphatase 1 regulatory subunit 21
Gene Aliases
EGK_05315, KLRAQ1
Chromosome Location
Chr.13: 60230960 - 60323275 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU332921.1 - - 2414 146
JU474323.1 - - 2372 146
JV706950.1 - - 2436 146

View Full Product Details