Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RDH10
Entrez Gene ID
695791
Gene Name
retinol dehydrogenase 10
Gene Aliases
-
Chromosome Location
Chr.8: 73801328 - 73829371 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266830.1 NP_001253759.1 - 658 118
GenBank mRNA JU474464.1 - - 706 118
JV635980.1 - - 658 118

View Full Product Details