Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM71D
Entrez Gene ID
713472
Gene Name
family with sequence similarity 71 member D
Gene Aliases
EGK_18300
Chromosome Location
Chr.7: 129318813 - 129359310 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194896.2 NP_001181825.1 - 907 91
XM_015143939.2 XP_014999425.2 - 1489 91
XM_015143940.2 XP_014999426.2 - 1407 91
XM_015143941.2 XP_014999427.2 - 1488 91
XM_015143942.2 XP_014999428.1 - 1482 91
XM_015143944.2 XP_014999430.2 - 828 91
XM_028850461.1 XP_028706294.1 - 1482 91
XM_028850462.1 XP_028706295.1 - 1482 91

View Full Product Details