Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP46
Entrez Gene ID
719561
Gene Name
cilia and flagella associated protein 46
Gene Aliases
C10orf93, C9H10orf93
Chromosome Location
Chr.9: 133393534 - 133407052 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190957.3 NP_001177886.3 - 637 113

View Full Product Details