Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTWD2
Entrez Gene ID
697989
Gene Name
DTW domain containing 2
Gene Aliases
-
Chromosome Location
Chr.6: 116479424 - 116637166 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261379.1 NP_001248308.1 - 427 117
XM_028849122.1 XP_028704955.1 - 496 117
XM_028849123.1 XP_028704956.1 - 281 117
GenBank mRNA JU323059.1 - - 427 117
JV649066.1 - - 211 117

View Full Product Details