Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC12
Entrez Gene ID
711072
Gene Name
zinc finger CCHC-type containing 12
Gene Aliases
EGK_20857
Chromosome Location
Chr.X: 115092448 - 115095619 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261787.1 NP_001248716.1 - 367 164
GenBank mRNA JU333215.1 - - 367 164
JV607733.1 - - 367 164

View Full Product Details