Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XG
Entrez Gene ID
705377
Gene Name
Xg glycoprotein (Xg blood group)
Gene Aliases
-
Chromosome Location
Chr.X: 2334247 - 2398610 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086077.4 XP_001086077.1 - 295 110
XM_028842380.1 XP_028698213.1 - 296 110
XM_028842381.1 XP_028698214.1 - 296 110
GenBank mRNA JU492878.1 - - 261 110

View Full Product Details