Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT79
Entrez Gene ID
700188
Gene Name
keratin 79
Gene Aliases
-
Chromosome Location
Chr.11: 52386538 - 52399464 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099606.3 XP_001099606.2 - 757 146

View Full Product Details