Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NME9
Entrez Gene ID
716434
Gene Name
NME/NM23 family member 9
Gene Aliases
TXNDC6
Chromosome Location
Chr.2: 40456422 - 40486228 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001114574.4 XP_001114574.3 - 652 127

View Full Product Details