Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCHF3
Entrez Gene ID
703693
Gene Name
membrane associated ring-CH-type finger 3
Gene Aliases
EGK_16802, MARCH3
Chromosome Location
Chr.6: 124356447 - 124504082 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266413.1 NP_001253342.1 - 441 112
GenBank mRNA JU320625.1 - - 279 112
JU472595.1 - - 442 112
JV046120.1 - - 441 112

View Full Product Details