Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83C
Entrez Gene ID
705662
Gene Name
family with sequence similarity 83 member C
Gene Aliases
-
Chromosome Location
Chr.10: 70495161 - 70501371 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194151.3 NP_001181080.2 - 930 136

View Full Product Details