Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMTM4
Entrez Gene ID
695709
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
-
Chromosome Location
Chr.20: 53787881 - 53869415 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015126334.2 XP_014981820.1 - 672 151
GenBank mRNA JU339176.1 - - 474 151

View Full Product Details