Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMTM8
Entrez Gene ID
704234
Gene Name
CKLF like MARVEL transmembrane domain containing 8
Gene Aliases
EGK_12170
Chromosome Location
Chr.2: 15343310 - 15473785 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097523.4 XP_001097523.1 - 438 111
GenBank mRNA JU326855.1 - - 59 111

View Full Product Details