Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS12
Entrez Gene ID
693778
Gene Name
transmembrane serine protease 12
Gene Aliases
EGK_03644
Chromosome Location
Chr.11: 50475736 - 50521399 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015151717.2 XP_015007203.1 - 902 135
XM_015151718.2 XP_015007204.1 - 846 135

View Full Product Details