Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MDGA2
Entrez Gene ID
703237
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
MAMDC1
Chromosome Location
Chr.7: 109043622 - 109874635 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097544.4 XP_001097544.1 - 2772 76
XM_015143645.2 XP_014999131.1 - 2410 76
XM_028851533.1 XP_028707366.1 - 2418 76
GenBank mRNA JU339561.1 - - 2459 76

View Full Product Details