Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIMC1
Entrez Gene ID
701880
Gene Name
SUMO interacting motifs containing 1
Gene Aliases
C5orf25
Chromosome Location
Chr.6: 173693606 - 173785545 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015141460.2 XP_014996946.2 - 2663 131
XM_015141462.2 XP_014996948.2 - 2498 131
GenBank mRNA JU325820.1 - - 974 131
JV060448.1 - - 988 131
JV714375.1 - - 989 131

View Full Product Details