Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC178
Entrez Gene ID
707566
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18H18orf34
Chromosome Location
Chr.18: 49056623 - 49575271 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101213.4 XP_001101213.2 - 1923 116
XM_015121789.2 XP_014977275.2 - 1923 116
XM_015121790.2 XP_014977276.2 - 1854 116
XM_015121791.2 XP_014977277.2 - 2011 116
XM_015121792.2 XP_014977278.2 - 1863 116

View Full Product Details