The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001093383.4 | XP_001093383.1 | - | 1730 | 73 |
GenBank mRNA | JU483822.1 | - | - | 1298 | 73 |