Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCN15
Entrez Gene ID
721696
Gene Name
lipocalin 15
Gene Aliases
EGK_07178
Chromosome Location
Chr.15: 1462959 - 1468336 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194733.2 NP_001181662.1 - 434 149
XM_015116327.2 XP_014971813.1 - 952 149

View Full Product Details