Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CERS6
Entrez Gene ID
704953
Gene Name
ceramide synthase 6
Gene Aliases
LASS6
Chromosome Location
Chr.12: 55591204 - 55904228 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266123.2 NP_001253052.1 - 1110 117
GenBank mRNA JU475026.1 - - 1112 117

View Full Product Details