Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS11F
Entrez Gene ID
710088
Gene Name
transmembrane serine protease 11F
Gene Aliases
-
Chromosome Location
Chr.5: 65424375 - 65503441 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028848547.1 XP_028704380.1 - 510 136

View Full Product Details