Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AVPR1A
Entrez Gene ID
717041
Gene Name
arginine vasopressin receptor 1A
Gene Aliases
-
Chromosome Location
Chr.11: 62907436 - 62914278 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001116798.4 XP_001116798.3 - 2551 112

View Full Product Details