Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4N2
Entrez Gene ID
701881
Gene Name
olfactory receptor, family 4, subfamily N, member 2
Gene Aliases
-
Chromosome Location
Chr.7: 81857188 - 81876176 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193869.2 NP_001180798.1 - 840 131

View Full Product Details