Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPGN
Entrez Gene ID
702632
Gene Name
epithelial mitogen
Gene Aliases
EGK_15789
Chromosome Location
Chr.5: 59390828 - 59397847 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102155.4 XP_001102155.1 - 317 79

View Full Product Details