Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXB2
Entrez Gene ID
704969
Gene Name
forkhead box B2
Gene Aliases
-
Chromosome Location
Chr.15: 92417453 - 92419239 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100107.2 XP_001100107.1 - 1230 123

View Full Product Details